4-Bromo-3-methylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

A188395-100G A188395-10G A188395-1G A188395-500G A188395-25G A188395-5G
103846-688EA 6 USD
103846-688 76998-904 76998-906 76998-908 76998-910 77084-414 77084-416
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole

Caution: for Research Use Only

Formula: C₈H₉BrO
Boiling Pt: 235.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR