2-Bromo-6-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A177502-100G
A177502-10G
A177502-25G
A177502-500G
A177502-5G
76998-414EA
99
USD
76998-414
76998-416
76998-418
76998-420
76998-422
2-Bromo-6-fluoropyridine 98%
2-Bromo-6-fluoropyridine
Caution: for Research Use Only
Formula:
C₅H₃BrFN |
MDL Number:
MFCD03095258 CAS Number: 144100-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...