Nɑ-(tert-Butoxycarbonyl)-D-lysine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nɑ-Boc-D-lysine, Boc-D-Lys-OH
A176922-1G
A176922-25G
A176922-5G
76998-396EA
9
USD
76998-396
76998-398
76998-400
Nɑ-(tert-Butoxycarbonyl)-D-lysine 97%
Nalpha-(tert-Butoxycarbonyl)-D-lysine
Caution: for Research Use Only
Formula:
C₁₁H₂₂N₂O₄ MW: 246.31 g/mol |
MDL Number:
MFCD00076956 CAS Number: 106719-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...