N,O-Dimethylhydroxylamine hydrochloride 98%

Supplier: Ambeed
Warning

A163050-1000G A163050-100G A163050-500G A163050-25G
76997-898EA 96 USD
76997-898 76997-900 76997-902 77320-022
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride

Caution: for Research Use Only

Formula: C₂H₈ClNO
MW: 97.55 g/mol
MDL Number: MFCD00012485
CAS Number: 6638-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR