Rosiglitazone 98%

Supplier: Ambeed
Warning

A158185-1MG A158185-25G A158185-50MG A158185-1G
103805-102EA 32 USD
103805-102 103843-310 76997-654 76997-656 76997-658 77319-162
Rosiglitazone 98%
Rosiglitazone

Caution: for Research Use Only

Formula: C₁₈H₁₉N₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00871760
CAS Number: 122320-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR