(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 98%

Supplier: Ambeed
Warning

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

A150620-100G A150620-10G A150620-500G A150620-1G A150620-25G A150620-5G A150620-1000G
103804-188EA 9 USD
103804-188 103842-324 76997-320 76997-322 76997-324 77299-818 77317-852 77317-854 77600-136
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 98%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

Caution: for Research Use Only

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Boiling Pt: 665.7 °C (760 mm Hg)
Melting Pt: 224…228 °C
Flash Pt: 449.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00233866
CAS Number: 161265-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR