Boc-L-3-benzothienylalanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-β-(3-benzothienyl)-Ala-OH, (S)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid
A103940-1G
A103940-250MG
A103940-5G
A103940-25G
76994-702EA
73
USD
76994-702
76994-704
76994-706
77880-841
Boc-L-3-benzothienylalanine 95%
Boc-L-3-benzothienylalanine
Caution: for Research Use Only
Formula:
C₁₆H₁₉NO₄S |
MDL Number:
MFCD00237538 CAS Number: 154902-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...