3,5-Difluoropyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoropicolinonitrile, 2-Cyano-3,5-difluoropyridine

A101495-100G A101495-10G A101495-25G A101495-1G A101495-5G A101495-500G
103796-002EA 8 USD
103796-002 103821-686 76994-498 76994-500 76994-502 77297-132 77297-134 77670-466
3,5-Difluoropyridine-2-carbonitrile 97%
3,5-Difluoropyridine-2-carbonitrile

Caution: for Research Use Only

Formula: C₆H₂F₂N₂
Boiling Pt: 187.8 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD03788759
CAS Number: 298709-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR