3,5-Difluoropyridine-2-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Difluoropicolinonitrile, 2-Cyano-3,5-difluoropyridine
A101495-100G
A101495-10G
A101495-25G
A101495-1G
A101495-5G
A101495-500G
103796-002EA
8
USD
103796-002
103821-686
76994-498
76994-500
76994-502
77297-132
77297-134
77670-466
3,5-Difluoropyridine-2-carbonitrile 97%
3,5-Difluoropyridine-2-carbonitrile
Caution: for Research Use Only
Formula:
C₆H₂F₂N₂ Boiling Pt: 187.8 °C (760 mm Hg) Melting Pt: 30…34 °C Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788759 CAS Number: 298709-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...