Human Serum Albumin (HSA) Solution (20%), 50 ml
Supplier: GeminiBio
800-121-050
76994-318EA 133.31 USD
76994-318
Human Serum Albumin (HSA) Solution (20%), 50 ml
Sera (Serum)
This diagnostic-grade product is an aqueous solution containing 20% human albumin (w/v). It is prepared by cold-alcohol fractionation from pooled human plasma obtained from venous blood. The USP Pharmacopeia requires that all fractionators add sodium caprylate and sodium tryptophanate as a preservative.
Delivery information: Ships on dry ice. Store frozen at −20 to −10 °C.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.