Human Serum Albumin (HSA) Powder-Low Endotoxin, 5 g
Supplier: GeminiBio
800-126P-005
76994-312EA 324.69 USD
76994-312
Human Serum Albumin (HSA) Powder-Low Endotoxin, 5 g
Sera (Serum)
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilized. MW ~66kDa. Endotoxin ≤2 EU/mg.
Delivery information: Ships at ambient. Store refrigerated at 2 to 8 °C
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.