PTS Panels® Multi-Chemistry Controls (Level 1 and 2)
Supplier: PTS Diagnostics
721
76994-102EA 77.1 USD
76994-102
PTS Panels® Multi-Chemistry Controls (Level 1 and 2)
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels Multi-Chemistry controls are for professional use with the CardioChek PA and CardioChek Plus analyzers and PTS Panels test strips as quality control materials to estimate precision and to detect systematic analytical deviations of your test system.
  • For professional CardioChek analyzers and PTS panels test strips
  • Liquid Volume
  • Room temperature storage until expiration or up to 10 months once opened
  • Opened or unopened product can be stored refrigerated until expiration
PTS Panels Multi-Chemistry Controls are for professional use with the CardioChek PA and CardioChek Plus analyzers and PTS Panels test strips as quality control materials to estimate precision and to detect systematic analytical deviations of your test system. The multi-chemistry controls, which are provided in two levels, contain cholesterol, triglycerides, and glucose.
Providers spend less time testing and more time counseling patients face-to-face during the same office visit for that 'teachable moment'. The CardioChek test system is CLIA-waived, meets NCEP guidelines for accuracy and precision, CRMLN-certfified, and reimbursable in most instances.
Certifications: CLIA-waived, FDA-cleared.