1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: (S,S)-Et-DUPHOS-Rh, (+)-1,2-BIS((2S,5S)-2,5-DIETHYLPHOSPHOLANO)BENZENE(CYCLOOCTADIENE)RHODIUM(L)TETRAFLUOROBORATE, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

A558457-100MG A558457-1G A558457-250MG
76991-744EA 191 USD
76991-744 76991-746 76991-748
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 98%
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

Caution: for Research Use Only

Formula: C₃₀H₄₆BF₄P₂Rh
MDL Number: MFCD01862465
CAS Number: 213343-64-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR