DL-Glutamine 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid

A228319-100G A228319-500G A228319-25G A228319-5G
103850-296EA 21 USD
103850-296 76987-226 76987-228 77303-828 77303-830
DL-Glutamine 97%
DL-Glutamine

Caution: for Research Use Only

Formula: C₅H₁₀N₂O₃
Boiling Pt: 445.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00065103
CAS Number: 6899-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR