D-(-)-Arabinose 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

A213618-100G A213618-10G A213618-500G A213618-25G
76986-810EA 31 USD
76986-810 76986-812 76986-814 77683-608
D-(-)-Arabinose 97%, Ambeed.Inc
D(-)-Arabinose

Caution: For research use only.

Formula: C₅H₁₀O₅
MDL Number: MFCD00135608
CAS Number: 10323-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR