4-Bromo-2-methoxythiazole 98%

Supplier: Ambeed
Warning

A199502-1G A199502-250MG
76986-398EA 39 USD
76986-398 76986-400
4-Bromo-2-methoxythiazole 98%
4-Bromo-2-methoxythiazole

Caution: for Research Use Only

Formula: C₄H₄BrNOS
MDL Number: MFCD07787374
CAS Number: 240816-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR