2-Chloro-3-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A151143-25G
A151143-5G
A151143-100G
76984-650EA
24
USD
76984-650
76984-652
77592-840
2-Chloro-3-fluoropyridine 98%
2-Chloro-3-fluoropyridine
Caution: for Research Use Only
Formula:
C₅H₃ClFN |
MDL Number:
MFCD03095302 CAS Number: 17282-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...