5-Amino-2,4-difluorophenol 95%

Supplier: Ambeed
Warning

Synonyms: 5-amino-2,4-difluorophenol

A129853-1G A129853-250MG
76983-618EA 13 USD
76983-618 76983-620
5-Amino-2,4-difluorophenol 95%
5-Amino-2,4-difluorophenol

Caution: for Research Use Only

Formula: C₆H₅F₂NO
MDL Number: MFCD04973718
CAS Number: 113512-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR