5-Carboxyfluorescein 97%

Supplier: Ambeed
Warning

Synonyms: Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate

A117509-100MG A117509-1MG A117509-250MG A117509-25G A117509-1G A117509-25MG A117509-5G
103792-054EA 13 USD
103792-054 103798-904 103799-298 103824-792 76983-050 76983-052 76983-054 76983-056 77297-542 77297-544 77297-546
5-Carboxyfluorescein 97%
5-Carboxyfluorescein

Caution: for Research Use Only

Formula: C₂₁H₁₂O₇.xH₂O
Boiling Pt: 725.2 °C (760 mm Hg)
Flash Pt: 266.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036874
CAS Number: 76823-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR