5-Carboxyfluorescein 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate
A117509-100MG
A117509-1MG
A117509-250MG
A117509-25G
A117509-1G
A117509-25MG
A117509-5G
103792-054EA
13
USD
103792-054
103798-904
103799-298
103824-792
76983-050
76983-052
76983-054
76983-056
77297-542
77297-544
77297-546
5-Carboxyfluorescein 97%
5-Carboxyfluorescein
Caution: for Research Use Only
Formula:
C₂₁H₁₂O₇.xH₂O Boiling Pt: 725.2 °C (760 mm Hg) Flash Pt: 266.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...