p-Toluenesulfonyl isocyanate 99%

Supplier: Ambeed
Danger

Synonyms: Tosyl isocyanate, 4-Isocyanatosulphonyltoluene, 4-Isocyanatosulfonyltoluene

A404059-25G
76979-100EA 6.37 USD
76979-100
p-Toluenesulfonyl isocyanate 99%
p-Toluenesulphonyl isocyanate

Caution: for Research Use Only

Formula: C₈H₇NO₃S
MW: 197.21 g/mol
MDL Number: MFCD00002030
CAS Number: 4083-64-1
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR