(R)-(+)-1-(1-Naphthyl)ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

A311072-25G A311072-100G A311072-10G A311072-5G A311072-500G
103835-838EA 7 USD
103835-838 76838-628 76977-902 76977-904 76977-906 77306-044 77672-588
(R)-(+)-1-(1-Naphthyl)ethylamine 98%
(R)-(+)-1-(1-Naphthyl)ethylamine

Caution: For research use only

Formula: C₁₂H₁₃N
Boiling Pt: 289.9 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064114
CAS Number: 3886-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR