Uridine 5′-monophosphate disodium salt 98%

Supplier: Ambeed
Warning

Synonyms: U 5′-P, Disodium uridine-5-monophosphate, Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate, UMP, 5′-UMP-Na2, di-Sodium uridine-5-monophosphate, 5'-UMP 2Na, 5'-Uridylic acid disodium salt, Uridylic acid disodium salt, 5'-UMP disodium salt, Disodium UMP

A260727-25G A260727-25MG A260727-5G A260727-100G A260727-500G A260727-1000G
76976-942EA 16 USD
76976-942 76976-944 76976-946 77601-098 77672-328 77672-326
Uridine 5′-monophosphate disodium salt 98%
Uridine 5′-monophosphate disodium salt

Caution: for Research Use Only

Formula: C₉H₁₁N₂Na₂O₉P
MW: 368.15 g/mol
MDL Number: MFCD00006525
CAS Number: 3387-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR