3,5,7,4'-Tetrahydroxy-3'-methoxyflavone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3'-Methoxy-3,4',5,7-tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-chromen-4-one, Isorhamnetin, 3'-Methylquercetin, 3-Methylquercetin
A189385-10MG
A189385-1MG
A189385-5MG
A189385-1G
A189385-250MG
A189385-50MG
A189385-100MG
76974-804EA
39
USD
76974-804
76974-806
76974-808
77671-684
77671-686
77671-688
77881-412
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone 97%
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Caution: for Research Use Only
Formula:
C₁₆H₁₂O₇ MW: 316.27 g/mol |
MDL Number:
MFCD00017310 CAS Number: 480-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...