2,6-Dichloropyridine-4-methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine
A185199-1G
A185199-250MG
A185199-5G
76974-576EA
6.37
USD
76974-576
76974-578
76974-580
76974-582
2,6-Dichloropyridine-4-methanol 97%
2,6-Dichloropyridine-4-methanol
Caution: for Research Use Only
Formula:
C₆H₅Cl₂NO |
MDL Number:
MFCD00052638 CAS Number: 101990-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...