6-Chloro-7-deazapurine 98%

Supplier: Ambeed
Warning

A145875-100G A145875-10G A145875-1G A145875-25G A145875-500G A145875-5G
76972-832EA 83 USD
76972-832 76972-834 76972-836 76972-838 76972-840 76972-842
6-Chloro-7-deazapurine 98%
6-Chloro-7-deazapurine

Caution: for Research Use Only

Formula: C₆H₄ClN₃
MDL Number: MFCD01686865
CAS Number: 3680-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR