2-chloro-5-iodopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-iodopyrimidine

A128084-100G A128084-10G A128084-250MG A128084-1G
103801-064EA 7 USD
103801-064 103831-940 76971-946 76971-948 76971-950 77313-350
2-chloro-5-iodopyrimidine 98%
2-chloro-5-iodopyrimidine

Caution: for Research Use Only

Formula: C₄H₂ClIN₂
Boiling Pt: 317.8 °C (760 mm Hg)
Melting Pt: 128…131 °C
Flash Pt: 146 °C
Storage Temperature: Freezer
MDL Number: MFCD06200210
CAS Number: 32779-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR