N-Phthaloylglycine 98%

Supplier: Ambeed
Warning

Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

A111911-100G A111911-10G A111911-1G A111911-25G A111911-500G A111911-5G A111911-1000G
76970-870EA 42 USD
76970-870 76970-872 76970-874 76970-876 76970-878 76970-880 77880-900
N-Phthaloylglycine 98%
N-Phthaloylglycine

Caution: for Research Use Only

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD00005900
CAS Number: 4702-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR