L-Chg-OMe·HCI 95%

Supplier: Ambeed
Warning

Synonyms: H-Chg-OMe·HCl, H-Chg-OMe.HCl

A105044-5G A105044-25G A105044-1G A105044-10G
76970-376EA 12 USD
76970-376 77565-830 77592-116 77880-850
L-Chg-OMe·HCI 95%
L-Chg-OMe·HCI

Caution: For research use only.

Formula: C₉H₁₇NO₂·HCl
MW: 207.74 g/mol
MDL Number: MFCD00797545
CAS Number: 14328-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR