Ethyl-4-iodobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

A104892-100G A104892-10G A104892-25G A104892-500G A104892-5G
76970-354EA 37 USD
76970-354 76970-356 76970-358 76970-360 76970-362
Ethyl-4-iodobenzoate 98%
Ethyl 4-iodobenzoate

Caution: for Research Use Only

Formula: C₉H₉IO₂
MDL Number: MFCD00017344
CAS Number: 51934-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR