Anti-Cyclin D1 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85171-100
76967-526EA 504.6 USD
76967-526
Anti-Cyclin D1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Cyclin D1 for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: Cyclin D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR