Anti-ICAM3 Rabbit Recombinant Antibody [clone: ICAM3/2873R]

Supplier: ANTIBODIES.COM LLC
A248913-100
76967-432EA 555.5 USD
76967-432
Anti-ICAM3 Rabbit Recombinant Antibody [clone: ICAM3/2873R]
Antibodies
Recombinant rabbit monoclonal [ICAM3/2873R] antibody to ICAM3 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: ICAM3
Clonality: Recombinant
Clone: ICAM3/2873R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR