Anti-ASCL1 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85121-100
76964-488EA 504.6 USD
76964-488
Anti-ASCL1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ASCL1 for ELISA and WB with samples derived from Mouse and Rat.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: ASCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR