Anti-Estrogen Receptor Mouse Monoclonal Antibody [clone: ER505]

Supplier: ANTIBODIES.COM LLC
A251656-100
76963-332EA 555.5 USD
76963-332
Anti-Estrogen Receptor Mouse Monoclonal Antibody [clone: ER505]
Antibodies
Mouse monoclonal [ER505] antibody to Estrogen Receptor for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: Estrogen Receptor
Clonality: Monoclonal
Clone: ER505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR