Anti-ARHGDIG Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84853-100
76962-548EA 504.6 USD
76962-548
Anti-ARHGDIG Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ARHGDIG for ELISA, WB and IHC with samples derived from Human.

  • Validated Applications: ELISA, WB, IH

Type: Primary
Antigen: ARHGDIG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR