Anti-Nprl3 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85003-100
76960-974EA 504.6 USD
76960-974
Anti-Nprl3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to NPRL3 for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: Nprl3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR