Anti-Ephrin B2 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84855-100
76958-178EA 504.6 USD
76958-178
Anti-Ephrin B2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Ephrin B2 for ELISA and WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: Ephrin B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR