Anti-Emerin Mouse Monoclonal Antibody [clone: EMD/2168]

Supplier: ANTIBODIES.COM LLC
A248426-100
76957-404EA 555.5 USD
76957-404
Anti-Emerin Mouse Monoclonal Antibody [clone: EMD/2168]
Antibodies
Mouse monoclonal [EMD/2168] antibody to Emerin for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Emerin
Clonality: Monoclonal
Clone: EMD/2168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR