Anti-Ulk3 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A85002-100
76956-924EA 504.6 USD
76956-924
Anti-Ulk3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ULK3 for ELISA and IHC with samples derived from Human.

  • Validated Applications: ELISA, IH

Type: Primary
Antigen: Ulk3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR