Anti-CD68 Mouse Monoclonal Antibody [Clone: C68/2709]

Supplier: ANTIBODIES.COM LLC
A253946-100
76953-212EA 555.5 USD
76953-212
Anti-CD68 Mouse Monoclonal Antibody [Clone: C68/2709]
Antibodies
Mouse monoclonal [C68/2709] antibody to CD68 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 0.25 to 0.5 µg/ml

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: C68/2709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR