Anti-ALDH9A1 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84830-100
76952-562EA 504.6 USD
76952-562
Anti-ALDH9A1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ALDH9A1 for ELISA, WB and IHC with samples derived from Human.

  • Validated Applications: ELISA, WB, IH

Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR