Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/2145]

Supplier: ANTIBODIES.COM LLC
A252262-100
76952-176EA 555.5 USD
76952-176
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/2145]
Antibodies
Mouse monoclonal [ITGB3/2145] antibody to Integrin beta 3 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: ITGB3/2145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR