Anti-POU2F2 Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84810-100
76950-632EA 504.6 USD
76950-632
Anti-POU2F2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to POU2F2 for ELISA and WB with samples derived from Human and Mouse.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: POU2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR