Plastic Staining Rack, Holds 24 Mopec Microscope Slides
Supplier: Mortech Manufacturing
SP234
76948-900EA 46.04 USD
76948-900
Plastic Staining Rack, Holds 24 Mopec Microscope Slides
Racks Staining Racks
Gray plastic staining rack.
  • Holds (24) 1×3" standard microscope slides
The handle is permanently attached, but permits closure of the 24-slide staining dish and easy insertion /removal of the microscope slides.