Thermal Transfer Ribbon for SlideMate, SP15, and PSLIM Printer
Supplier: Mortech Manufacturing
SP202
76948-870EA 214.29 USD
76948-870
Thermal Transfer Ribbon for SlideMate, SP15, and PSLIM Printer
Printer Ribbons
Black, hot foil tape.
  • Used for SlideMate, SP15, and PSLIM printers
  • 984 feet per roll
  • Resistant to all reagents and alcohols