Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 40 mm, Available in Multiple Colors
Supplier: Mortech Manufacturing
SP104-BL SP104-GN SP104-OR SP104-PK SP104-PR SP104-RD SP104W-BL SP104W-GN SP104-WH SP104W-OR SP104W-PK SP104W-PR SP104W-RD SP104W-WH SP104W-YE SP104-YE
76948-544EA 142.75 USD
76948-544 76948-546 76948-548 76948-550 76948-552 76948-554 76948-596 76948-598 76948-556 76948-600 76948-602 76948-604 76948-606 76948-608 76948-610 76948-558
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 40 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for leica (except CM1950 model), TBS, and tanner cryostats
  • 40 mm
  • Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white.