Circular Specimen Chuck 25 mm Kit for Leica, TBS, and Tanner Cryostats
Supplier: Mortech Manufacturing
SP102-KT
76948-576EA 1110.54 USD
76948-576
Circular Specimen Chuck 25 mm Kit for Leica, TBS, and Tanner Cryostats
Specimen Mounting Supplies
SP102-KT: (6) 25 mm, (2) 30 mm, and (2) 40 mm in different colors, an angular brush, and a teasing needle.
  • Round, aluminum specimen chucks
  • Suitable for leica (except CM1950 model), TBS, and tanner cryostats
  • Standard kit includes 10 common leica-type specimen chucks, an angular brush and a teasing needle