Specimen Chuck Stand, Stainless Steel
Supplier: Mopec
SP100
76948-494EA 191.1 USD
76948-494
Specimen Chuck Stand, Stainless Steel
Specimen Mounting Supplies
Stainless steel stand for specimen chucks. Can be used in chamber to add additional specimens in cryostat, or as a staging stand.
  • Stainless steel
  • Dimensions: 8⁷/₈×2³/₄×1¹/₄"
  • Can be used in chamber to add additional specimens in cryostat, or as a staging stand