Microscope Slides, Charged, Inkjet, 45° Corners
Supplier: Mortech Manufacturing
SL245-BL-BX SL245-BL-CS SL245-GR-BX SL245-GR-CS SL245-LI-BX SL245-LI-CS SL245-PE-BX SL245-PE-CS SL245-PK-BX SL245-PK-CS SL245-TN-BX SL245-TN-CS SL245-WH-BX SL245-WH-CS SL245-YL-BX SL245-YL-CS
76948-418BX 63.24 USD
76948-418 76948-420 76948-422 76948-424 76948-426 76948-428 76948-430 76948-432 76948-434 76948-436 76948-438 76948-440 76948-442 76948-444 76948-446 76948-448
Microscope Slides, Charged, Inkjet, 45° Corners
Slides Microscope Slides
High-quality, charged microscope slides with color coding.
  • For Inkjet
  • 1×3" slides, 1mm thickness
  • 100% guaranteed against sticking and contamination
  • 45 degree corners
  • Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac.
Packaging: The packaging options include 1 box of 72 slides and 1 case of 1440 slides.