Microscope Slides, Charged, 45° Corners
Supplier: Mortech Manufacturing
SL145-GR-BX SL145-GR-CS SL145-LI-BX SL145-LI-CS SL145-PE-BX SL145-PE-CS SL145-PK-BX SL145-PK-CS SL145-TN-BX SL145-TN-CS SL145-BL-BX SL145-BL-CS SL145-WH-BX SL145-WH-CS SL145-YL-BX SL145-YL-CS
76948-358BX 63.24 USD
76948-358 76948-360 76948-362 76948-364 76948-366 76948-368 76948-370 76948-372 76948-374 76948-376 76948-354 76948-356 76948-378 76948-380 76948-382 76948-384
Microscope Slides, Charged, 45° Corners
Slides Microscope Slides
High-quality, charged microscope slides with color coding.
  • 1×3" slides, 1 mm thickness
  • 100% guaranteed against sticking and contamination
  • 45 degree corners
  • Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac.
Packaging: The packaging options include 1 box of 72 slides and 1 case of 1440 slides.